5-Chlorooctafluoropentanoic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC408092-25G
PC408092-5G
APOSPC408092-25GEA
3990
DKK
APOSPC408092-25G
APOSPC408092-5G
5-Chlorooctafluoropentanoic acid 95%
5-Chlorooctafluoropentanoic acid
Formel:
C₅HClF₈O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01328318 CAS-nummer: 66443-79-6 UN nummer: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...