cis-tert-Butyl 3-(3-fluorophenyl)-aziridine-2-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC404513-1G PC404513-250MG
APOSPC404513-1GEA 3820 DKK
APOSPC404513-1G APOSPC404513-250MG
cis-tert-Butyl 3-(3-fluorophenyl)-aziridine-2-carboxylate
cis-tert-Butyl 3-(3-fluorophenyl)-aziridine-2-carboxylate
Formel: C₁₃H₁₆FNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26936315
CAS-nummer: 1980007-29-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR