3-(4-Fluorophenyl)piperidin-3-ol hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC402145-1G PC402145-250MG
APOSPC402145-1GEA 3960 DKK
APOSPC402145-1G APOSPC402145-250MG
3-(4-Fluorophenyl)piperidin-3-ol hydrochloride
3-(4-Fluorophenyl)piperidin-3-ol hydrochloride
Formel: C₁₁H₁₄FNO·ClH
Molekylvægt: 231.694 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11501328
CAS-nummer: 173447-91-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR