Ethyl 3-amino-2,4-dichloro-5-fluorobenzoate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,6-Dichloro-3-(ethoxycarbonyl)-5-fluoroaniline

PC400518-1G PC400518-250MG
APOSPC400518-1GEA 3830 DKK
APOSPC400518-1G APOSPC400518-250MG
Ethyl 3-amino-2,4-dichloro-5-fluorobenzoate
Ethyl 3-amino-2,4-dichloro-5-fluorobenzoate
Formel: C₉H₈Cl₂FNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11500226
CAS-nummer: 1089333-20-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR