1-(5-Chloropyridin-3-yl)-2,2,2-trifluoroethanone

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC400095-1G PC400095-250MG
APOSPC400095-1GEA 2180 DKK
APOSPC400095-1G APOSPC400095-250MG
1-(5-Chloropyridin-3-yl)-2,2,2-trifluoroethanone
1-(5-Chloropyridin-3-yl)-2,2,2-trifluoroethanone
Formel: C₇H₃ClF₃NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13189039
CAS-nummer: 1060802-11-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR