6-Amino-3-bromo-2-fluorophenol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Bromo-3-fluoro-2-hydroxyaniline

PC400012-1G PC400012-250MG
APOSPC400012-1GEA 4580 DKK
APOSPC400012-1G APOSPC400012-250MG
6-Amino-3-bromo-2-fluorophenol 95%
6-Amino-3-bromo-2-fluorophenol
Formel: C₆H₅BrFNO
Molekylvægt: 206.012 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26407502
CAS-nummer: 1807234-85-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR