2-[Bromo(difluoro)methyl]-4-nitro-1H-benzimidazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC32929-1G
PC32929-250MG
PC32929-5G
APOSPC32929-1GEA
1300
DKK
APOSPC32929-1G
APOSPC32929-250MG
APOSPC32929-5G
2-[Bromo(difluoro)methyl]-4-nitro-1H-benzimidazole
2-[Bromo(difluoro)methyl]-4-nitro-1H-benzimidazole
Formel:
C₈H₄BrF₂N₃O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD31922512 CAS-nummer: 2366994-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...