5-Bromo-2-[bromo(difluoro)methyl]-1H-benzimidazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC32918-1G PC32918-250MG PC32918-5G
APOSPC32918-1GEA 676 DKK
APOSPC32918-1G APOSPC32918-250MG APOSPC32918-5G
5-Bromo-2-[bromo(difluoro)methyl]-1H-benzimidazole
5-Bromo-2-[bromo(difluoro)methyl]-1H-benzimidazole
Formel: C₈H₄Br₂F₂N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31922501
CAS-nummer: 1353778-39-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR