Heptafluoroisopropylbenzene 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC3137-1G PC3137-5G
APOSPC3137-1GEA 1140 DKK
APOSPC3137-1G APOSPC3137-5G
Heptafluoroisopropylbenzene 98%
Heptafluoroisopropylbenzene
Formel: C₉H₅F₇
Kogepunkt: 127…128 °C
Smeltepunkt: –10 °C
Massefylde: 1,488 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02093306
CAS-nummer: 378-34-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR