3-Bromo-4-methyl-5-(trifluoromethyl)benzyl alcohol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC303600-1G PC303600-5G
APOSPC303600-1GEA 2360 DKK
APOSPC303600-1G APOSPC303600-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl alcohol
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl alcohol
Formel: C₉H₈BrF₃O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD32640173
CAS-nummer: 2385206-02-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR