5-Bromo-3-chloro-2-fluoroaniline 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC303461-1G PC303461-5G
APOSPC303461-1GEA 2310 DKK
APOSPC303461-1G APOSPC303461-5G
5-Bromo-3-chloro-2-fluoroaniline 97%
5-Bromo-3-chloro-2-fluoroaniline
Formel: C₆H₄BrClFN
Molekylvægt: 224.458 g/mol
Smeltepunkt: 40…50 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28143920
CAS-nummer: 1517200-74-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR