2,3,4-Trifluoro-5-methylbenzonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC303442-1G PC303442-5G
APOSPC303442-1GEA 2620 DKK
APOSPC303442-1G APOSPC303442-5G
2,3,4-Trifluoro-5-methylbenzonitrile
2,3,4-Trifluoro-5-methylbenzonitrile
Formel: C₈H₄F₃N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD32640073
CAS-nummer: 1533973-82-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR