3-Chloro-5-(trifluoromethoxy)benzoyl chloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC302947-10G PC302947-5G
APOSPC302947-10GEA 0 DKK
APOSPC302947-10G APOSPC302947-5G
3-Chloro-5-(trifluoromethoxy)benzoyl chloride
3-Chloro-5-(trifluoromethoxy)benzoyl chloride
Formel: C₈H₃Cl₂F₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06660307
CAS-nummer: 886503-20-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR