3-Fluoro-2-methylbenzamide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC302777-10G PC302777-5G PC302777-1G PC302777-25G
APOSPC302777-10GEA 1430 DKK
APOSPC302777-10G APOSPC302777-5G APOSPC302777-1G APOSPC302777-25G
3-Fluoro-2-methylbenzamide 98%
3-Fluoro-2-methylbenzamide
Formel: C₈H₈FNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06660276
CAS-nummer: 886502-05-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR