3'-Chloro-5'-fluoropropiophenone 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC302774-10G PC302774-5G PC302774-1G
APOSPC302774-10GEA 4130 DKK
APOSPC302774-10G APOSPC302774-5G APOSPC302774-1G
3'-Chloro-5'-fluoropropiophenone 98%
3'-Chloro-5'-fluoropropiophenone
Formel: C₉H₈ClFO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115836
CAS-nummer: 886496-66-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR