4-Chloro-3-fluoro-5-(trifluoromethyl)benzyl bromide 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC302329-1G PC302329-5G PC302329-10G
APOSPC302329-1GEA 1490 DKK
APOSPC302329-1G APOSPC302329-5G APOSPC302329-10G
4-Chloro-3-fluoro-5-(trifluoromethyl)benzyl bromide 97%
4-Chloro-3-fluoro-5-(trifluoromethyl)benzyl bromide
Formel: C₈H₄BrClF₄
Molekylvægt: 291.468 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD23795334
CAS-nummer: 1431329-80-4
UN nummer: 3261
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR