1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-5-carbonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC300998-1G PC300998-250MG
APOSPC300998-1GEA 3390 DKK
APOSPC300998-1G APOSPC300998-250MG
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-5-carbonitrile
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-5-carbonitrile
Formel: C₁₅H₇ClF₃N₃
Molekylvægt: 321.684 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31630105
CAS-nummer: 151387-94-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR