1-{[4-Bromo-3-(trifluoromethyl)phenyl]sulphonyl}-4-methylpiperazine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC300691-1G PC300691-5G
APOSPC300691-1GEA 1490 DKK
APOSPC300691-1G APOSPC300691-5G
1-{[4-Bromo-3-(trifluoromethyl)phenyl]sulphonyl}-4-methylpiperazine
1-{[4-Bromo-3-(trifluoromethyl)phenyl]sulphonyl}-4-methylpiperazine
Formel: C₁₂H₁₄BrF₃N₂O₂S
Molekylvægt: 387.216 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD19442764
CAS-nummer: 486422-42-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR