4-Bromo-2-fluorobenzyl fluoride 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC27931-1G PC27931-250MG
APOSPC27931-1GEA 4580 DKK
APOSPC27931-1G APOSPC27931-250MG
4-Bromo-2-fluorobenzyl fluoride 95%
4-Bromo-2-fluorobenzyl fluoride
Formel: C₇H₅BrF₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31664908
CAS-nummer: 2244086-30-2
UN nummer: 3265
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR