Perfluoro(2-iodo-2-methylpentane) 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1,1,2,2,3,3,5,5,5-Decafluoro-2-iodo-2-(trifluoromethyl)pentane, 2-Iodoperfluoro(2-methylpentane)

PC2151G-1G PC2151G-5G
APOSPC2151G-1GEA 436 DKK
APOSPC2151G-1G APOSPC2151G-5G
Perfluoro(2-iodo-2-methylpentane) 97%
Perfluoro(2-iodo-2-methylpentane)
Formel: C₆F₁₃I
Kogepunkt: 115 °C
Massefylde: 2,14 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00082619
CAS-nummer: 102780-88-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR