1-(2-Fluoro-4-nitrophenyl)-4-phenylpiperazine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC210083-1G PC210083-500MG
APOSPC210083-1GEA 5560 DKK
APOSPC210083-1G APOSPC210083-500MG
1-(2-Fluoro-4-nitrophenyl)-4-phenylpiperazine
1-(2-Fluoro-4-nitrophenyl)-4-phenylpiperazine
Formel: C₁₆H₁₆FN₃O₂
Molekylvægt: 301.316 g/mol
Smeltepunkt: 152…153 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28506332
CAS-nummer: 556801-40-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR