Methyl 3-(4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)-2-acetamidoprop-2-enoate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC210066-1G PC210066-500MG
APOSPC210066-1GEA 2120 DKK
APOSPC210066-1G APOSPC210066-500MG
Methyl 3-(4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)-2-acetamidoprop-2-enoate
Methyl 3-(4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)-2-acetamidoprop-2-enoate
Formel: C₁₈H₁₄ClF₃N₂O₄
Molekylvægt: 414.763 g/mol
Smeltepunkt: 179…180 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28127025
CAS-nummer: 1630103-73-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR