3,6-Difluoropyridin-2-amine 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC201381-100MG PC201381-250MG PC201381-1G
APOSPC201381-100MGEA 1070 DKK
APOSPC201381-100MG APOSPC201381-250MG APOSPC201381-1G
3,6-Difluoropyridin-2-amine 95+%
3,6-Difluoropyridin-2-amine
Formel: C₅H₄F₂N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13175493
CAS-nummer: 944799-22-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR