2,2,2-Trifluoro-1-(p-tolyl)ethanol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC201255-250MG
PC201255-1G
APOSPC201255-250MGEA
285
DKK
APOSPC201255-250MG
APOSPC201255-1G
2,2,2-Trifluoro-1-(p-tolyl)ethanol 98%
2,2,2-Trifluoro-1-(p-tolyl)ethanol
Formel:
C₉H₉F₃O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00233111 CAS-nummer: 446-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...