4-Fluoro-5-methoxyisoindoline hydrochloride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC201141-100MG PC201141-1G PC201141-250MG
APOSPC201141-100MGEA 1200 DKK
APOSPC201141-100MG APOSPC201141-1G APOSPC201141-250MG
4-Fluoro-5-methoxyisoindoline hydrochloride 97%
4-Fluoro-5-methoxyisoindoline hydrochloride
Formel: C₉H₁₀FNO·ClH
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD25542148
CAS-nummer: 1447606-44-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR