2-(3-Methyl-5-(trifluoromethyl)phenyl)acetic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC201074-1G PC201074-5G
APOSPC201074-1GEA 1250 DKK
APOSPC201074-1G APOSPC201074-5G
2-(3-Methyl-5-(trifluoromethyl)phenyl)acetic acid 97%
2-(3-Methyl-5-(trifluoromethyl)phenyl)acetic acid
Formel: C₁₀H₉F₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13194442
CAS-nummer: 1214352-00-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR