3-Fluorophenylboronic acid pinacol ester
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(3-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PC200195-10G
PC200195-1G
PC200195-5G
APOSPC200195-10GEA
330
DKK
APOSPC200195-10G
APOSPC200195-1G
APOSPC200195-5G
3-Fluorophenylboronic acid pinacol ester
3-Fluorophenylboronic acid pinacol ester
Formel:
C₁₂H₁₆BFO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD18375236 CAS-nummer: 936618-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...