Tetrafluorosuccinimide
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC1323-250MG
PC1323-1G
APOSPC1323-250MGEA
861
DKK
APOSPC1323-250MG
APOSPC1323-1G
Tetrafluorosuccinimide
Tetrafluorosuccinimide
Formel:
C₄HF₄NO₂ Kogepunkt: 176 °C Smeltepunkt: 66…67 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07784309 CAS-nummer: 377-33-3 UN nummer: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...