4-(Trifluoromethyl)benzal chloride 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: PTF-BAC, 4-(Dichloromethyl)benzotrifluoride

PC1305-25G PC1305-5G PC1305-1G
APOSPC1305-25GEA 5180 DKK
APOSPC1305-25G APOSPC1305-5G APOSPC1305-1G
4-(Trifluoromethyl)benzal chloride 95%
4-(Trifluoromethyl)benzal chloride
Formel: C₈H₅Cl₂F₃
Kogepunkt: 76 °C (8 mm Hg)
Smeltepunkt: –3 °C
Massefylde: 1,4 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00274358
CAS-nummer: 82510-98-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR