2-Amino-3-fluorobutanoic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-3-fluorobutyric acid

PC1076D-100MG PC1076D-500MG
APOSPC1076D-100MGEA 1490 DKK
APOSPC1076D-100MG APOSPC1076D-500MG
2-Amino-3-fluorobutanoic acid 97%
2-Amino-3-fluorobutanoic acid
Formel: C₄H₈FNO₂
Smeltepunkt: 186…187 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00077450
CAS-nummer: 50885-01-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR