3-Iodo-4-(trifluoromethyl)benzyl alcohol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-(Hydroxymethyl)-2-iodobenzotrifluoride, [3-Iodo-4-(trifluoromethyl)phenyl]methanol

PC10672-1G PC10672-5G PC10672-250MG
APOSPC10672-1GEA 1310 DKK
APOSPC10672-1G APOSPC10672-5G APOSPC10672-250MG
3-Iodo-4-(trifluoromethyl)benzyl alcohol
3-Iodo-4-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆F₃IO
Molekylvægt: 302.032 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18398175
CAS-nummer: 372120-52-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR