4-Hydroxynonafluorobiphenyl 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC10608-1G PC10608-5G
APOSPC10608-1GEA 676 DKK
APOSPC10608-1G APOSPC10608-5G
4-Hydroxynonafluorobiphenyl 97%
4-Hydroxynonafluorobiphenyl
Formel: C₁₂HF₉O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134468
CAS-nummer: 2894-87-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR