2-Fluoropropan-1-ol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC1014-1G PC1014-5G PC1014-250MG
APOSPC1014-1GEA 1430 DKK
APOSPC1014-1G APOSPC1014-5G APOSPC1014-250MG
2-Fluoropropan-1-ol 97%
2-Fluoropropan-1-ol
Formel: C₃H₇FO
Kogepunkt: 115 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07777178
CAS-nummer: 3824-87-1
UN nummer: 1987
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR