3-Amino-5-bromo-4-methylbenzotrifluoride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC0955-1G PC0955-5G
APOSPC0955-1GEA 2390 DKK
APOSPC0955-1G APOSPC0955-5G
3-Amino-5-bromo-4-methylbenzotrifluoride 97%
3-Amino-5-bromo-4-methylbenzotrifluoride
Formel: C₈H₇BrF₃N
Molekylvægt: 254.047 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631688
CAS-nummer: 232267-31-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR