3,5-Bis(trifluoromethyl)phenylacetyl chloride 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC0707-5G
PC0707-10G
APOSPC0707-5GEA
654
DKK
APOSPC0707-5G
APOSPC0707-10G
3,5-Bis(trifluoromethyl)phenylacetyl chloride 97%
3,5-Bis(trifluoromethyl)phenylacetyl chloride
Formel:
C₁₀H₅ClF₆O Molekylvægt: 290.59 g/mol Kogepunkt: 103 °C (10 mm Hg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03788501 CAS-nummer: 174083-39-7 UN nummer: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...