4-Chloro-3-(trifluoromethyl)cinnamic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-[4-Chloro-3-(trifluoromethyl)phenyl]prop-2-enoic acid, 3-[4-Chloro-3-(trifluoromethyl)phenyl]acrylic acid

PC0196-1G PC0196-5G PC0196-10G
APOSPC0196-1GEA 273 DKK
APOSPC0196-1G APOSPC0196-5G APOSPC0196-10G
4-Chloro-3-(trifluoromethyl)cinnamic acid 97%
4-Chloro-3-(trifluoromethyl)cinnamic acid
Formel: C₁₀H₆ClF₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631407
CAS-nummer: 58537-09-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR