3-Hydroxyisonicotinamide 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR965079-1G OR965079-250MG OR965079-5G
APOSOR965079-1GEA 1140 DKK
APOSOR965079-1G APOSOR965079-250MG APOSOR965079-5G
3-Hydroxyisonicotinamide 95%
3-Hydroxyisonicotinamide
Formel: C₆H₆N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13659443
CAS-nummer: 10128-73-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR