2-Chloro-4-methoxypyridine-3-carbaldehyde 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR959688-1G
OR959688-250MG
APOSOR959688-1GEA
355
DKK
APOSOR959688-1G
APOSOR959688-250MG
2-Chloro-4-methoxypyridine-3-carbaldehyde 98%
2-Chloro-4-methoxypyridine-3-carbaldehyde
Formel:
C₇H₆ClNO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD15474989 CAS-nummer: 905563-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...