6-(Ethylamino)pyridine-3-carbonitrile 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR959603-10G OR959603-1G OR959603-5G
APOSOR959603-10GEA 5670 DKK
APOSOR959603-10G APOSOR959603-1G APOSOR959603-5G
6-(Ethylamino)pyridine-3-carbonitrile 97%
6-(Ethylamino)pyridine-3-carbonitrile
Formel: C₈H₉N₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09940529
CAS-nummer: 1016813-34-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR