2-(Isobutylcarbonyl)pyridine 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR959515-10G
OR959515-1G
OR959515-5G
APOSOR959515-10GEA
6980
DKK
APOSOR959515-10G
APOSOR959515-1G
APOSOR959515-5G
2-(Isobutylcarbonyl)pyridine 98%
2-(Isobutylcarbonyl)pyridine
Formel:
C₁₀H₁₃NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07699213 CAS-nummer: 6952-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...