Ethyl 2,3-dihydrobenzofuran-5-carboxylate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR959377-1G OR959377-5G
APOSOR959377-1GEA 2180 DKK
APOSOR959377-1G APOSOR959377-5G
Ethyl 2,3-dihydrobenzofuran-5-carboxylate 95%
Ethyl 2,3-dihydrobenzofuran-5-carboxylate
Formel: C₁₁H₁₂O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06204418
CAS-nummer: 83751-12-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR