(R)-b-Amino-3-bromo-benzeneethanol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (R)-2-Amino-2-(3-bromophenyl)ethanol

OR959135-1G OR959135-250MG OR959135-500MG
APOSOR959135-1GEA 6540 DKK
APOSOR959135-1G APOSOR959135-250MG APOSOR959135-500MG
(R)-b-Amino-3-bromo-benzeneethanol 97%
(R)-b-Amino-3-bromo-benzeneethanol
Formel: C₈H₁₀BrNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09253670
CAS-nummer: 209963-04-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR