(6-Chloropyridin-2-yl)methanamine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR959050-1G OR959050-250MG OR959050-5G
APOSOR959050-1GEA 1780 DKK
APOSOR959050-1G APOSOR959050-250MG APOSOR959050-5G
(6-Chloropyridin-2-yl)methanamine 95%
(6-Chloropyridin-2-yl)methanamine
Formel: C₆H₇ClN₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11046128
CAS-nummer: 188637-75-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR