3-Chloro-N-methoxy-N-methylbenzamide 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR957161-1G OR957161-5G
APOSOR957161-1GEA 162 DKK
APOSOR957161-1G APOSOR957161-5G
3-Chloro-N-methoxy-N-methylbenzamide 95%
3-Chloro-N-methoxy-N-methylbenzamide
Formel: C₉H₁₀ClNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02684307
CAS-nummer: 145959-21-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR