Ethyl 6-methylpyridazine-3-carboxylate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR953692-1G OR953692-250MG OR953692-5G
APOSOR953692-1GEA 2240 DKK
APOSOR953692-1G APOSOR953692-250MG APOSOR953692-5G
Ethyl 6-methylpyridazine-3-carboxylate 98%
Ethyl 6-methylpyridazine-3-carboxylate
Formel: C₈H₁₀N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14584681
CAS-nummer: 64210-57-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR