7-Nitro-3,4-dihydro-2H-1,5-benzodioxepine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR953455-1G OR953455-5G
APOSOR953455-1GEA 1260 DKK
APOSOR953455-1G APOSOR953455-5G
7-Nitro-3,4-dihydro-2H-1,5-benzodioxepine 95%
7-Nitro-3,4-dihydro-2H-1,5-benzodioxepine
Formel: C₉H₉NO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00508323
CAS-nummer: 78288-94-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR