Ethyl-2-tetrahydrofurfuroyl-acetate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethyl 3-oxo-3-(tetrahydrofuran-2-yl)propanoate

OR953328-1G OR953328-5G
APOSOR953328-1GEA 436 DKK
APOSOR953328-1G APOSOR953328-5G
Ethyl-2-tetrahydrofurfuroyl-acetate 95%
Ethyl-2-tetrahydrofurfuroyl-acetate
Formel: C₉H₁₄O₄
Molekylvægt: 186.205 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06410652
CAS-nummer: 887411-85-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR