5-Chloro-2-methoxybenzyl chloride 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Chloro-2-(chloromethyl)-1-methoxybenzene

OR952931-250MG OR952931-1G
APOSOR952931-250MGEA 1210 DKK
APOSOR952931-250MG APOSOR952931-1G
5-Chloro-2-methoxybenzyl chloride 98%
5-Chloro-2-methoxybenzyl chloride
Formel: C₈H₈Cl₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02182262
CAS-nummer: 7035-11-2
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR