Methyl 7-aminoheptanoate hydrochloride 98%
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
OR951446-25G OR951446-100G
APOSOR951446-25GEA 480 DKK
APOSOR951446-25G APOSOR951446-100G
Methyl 7-aminoheptanoate hydrochloride 98%
Methyl 7-aminoheptanoate hydrochloride
Formel: C₈H₁₇NO₂·ClH
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09029389
CAS-nummer: 17994-94-4