4-Methyl valeryl piperidine thioamide 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR951082-1G
OR951082-250MG
APOSOR951082-1GEA
6320
DKK
APOSOR951082-1G
APOSOR951082-250MG
4-Methyl valeryl piperidine thioamide 95%
4-Methyl valeryl piperidine thioamide
Formel:
C₁₁H₂₁NS Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02179413 CAS-nummer: 289677-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...